accessibility menu, dialog, popup

UserName

CAS RN 32387-21-6

IUPAC Name: 1-methyl-1H-indole-3-carboxylic acid
Synonyms: 1-methylindole-3-carboxylic acid
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: HVRCLXXJIQTXHC-UHFFFAOYSA-N
SMILES: CN1C=C(C(O)=O)C2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results