accessibility menu, dialog, popup

UserName

Se(S)NH2OHOH3C

CAS RN 3211-76-5

IUPAC Name: (2S)-2-amino-4-(methylselanyl)butanoic acid
Synonyms: L-selenomethionine
Molecular Weight (g/mol): 196.12
Molecular Formula: C5H11NO2Se
InChi Key: RJFAYQIBOAGBLC-BYPYZUCNSA-N
SMILES: C[Se]CC[C@H](N)C(O)=O
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (6)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)