missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Selenomethionine, Calbiochem™,

Catalog No. 561505250MG
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
561505250MG 250 mg
5615051GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 561505250MG Supplier MilliporeSigma™ Supplier No. 561505250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

Format Glass Bottle

Specifications

CAS 3211-76-5
Color White
Form Solid
Molecular Formula C5H11NO2Se
MDL Number MFCD00037210
Synonym l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid
InChI Key RJFAYQIBOAGBLC-BYPYZUCNSA-N
SMILES C[Se]CC[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-4-(methylselanyl)butanoic acid
Molecular Weight (g/mol) 196.12
PubChem CID 105024
ChEBI CHEBI:62621
Quantity 250 mg
Percent Purity ≥98% (HPLC)
Show More Show Less