accessibility menu, dialog, popup

UserName

CAS RN 3204-73-7

CH3OOOOH

CAS RN 3204-73-7

IUPAC Name: methyl 4-(2-hydroxyethoxy)benzoate
Synonyms: methyl 4-(2-hydroxyethoxy)benzoate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: VFBYWNSASHHPPA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OCCO)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results