missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(2-Hydroxyethoxy)benzoate 98.0+%, TCI America™

Catalog No. H08205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08205G 5 g
H082025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08205G Supplier TCI America Supplier No. H08205G

Chemical Identifiers

CAS 3204-73-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00075872
InChI Key VFBYWNSASHHPPA-UHFFFAOYSA-N
Synonym 4-(2-Hydroxyethoxy)benzoic Acid Methyl Ester
PubChem CID 76670
IUPAC Name methyl 4-(2-hydroxyethoxy)benzoate
SMILES COC(=O)C1=CC=C(OCCO)C=C1

Specifications

Melting Point 66°C
Color White
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(2-Hydroxyethoxy)benzoate