accessibility menu, dialog, popup

UserName

CAS RN 31645-35-9

IUPAC Name: (anthracen-9-yl)methyl 2-methylprop-2-enoate
Synonyms: anthracen-9-ylmethyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 276.34
Molecular Formula: C19H16O2
InChi Key: MJYSISMEPNOHEG-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC1=C2C=CC=CC2=CC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results