missing translation for 'onlineSavingsMsg'
Learn More

9-Anthrylmethyl Methacrylate 98.0+%, TCI America™

Catalog No. A26451G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26451G Supplier TCI America Supplier No. A26451G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31645-35-9
Molecular Formula C19H16O2
Molecular Weight (g/mol) 276.335
MDL Number MFCD00673247
InChI Key MJYSISMEPNOHEG-UHFFFAOYSA-N
Synonym Methacrylic Acid 9-Anthrylmethyl Ester
PubChem CID 3618754
IUPAC Name anthracen-9-ylmethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1=C2C=CC=CC2=CC3=CC=CC=C31

Specifications

Melting Point 88°C
Color Yellow
Quantity 1 g
Formula Weight 276.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Anthrylmethyl Methacrylate