CAS RN 31618-90-3
IUPAC Name:
diethyl [(4-methylbenzenesulfonyl)oxy]methanephosphonate
Synonyms:
diethyl [(4-methylbenzenesulfonyl)oxy]methanephosphonate
Molecular Weight (g/mol):
322.31
Molecular Formula:
C12H19O6PS
InChi Key:
UOEFFQWLRUBDME-UHFFFAOYSA-N
SMILES:
CCOP(=O)(COS(=O)(=O)C1=CC=C(C)C=C1)OCC
1
–
1
of
1
results
1
Diethyl (p-Toluenesulfonyloxymethyl)phosphonate 97.0+%, TCI America™
CAS: 31618-90-3 Molecular Formula: C12H19O6PS Molecular Weight (g/mol): 322.312 MDL Number: MFCD03412078 InChI Key: UOEFFQWLRUBDME-UHFFFAOYSA-N Synonym: (p-Toluenesulfonyloxymethyl)phosphonic Acid Diethyl Ester, (Diethoxyphosphoryl)methyl p-Toluenesulfonate, p-Toluenesulfonic Acid (Diethoxyphosphoryl)methyl Ester PubChem CID: 4193599 IUPAC Name: diethoxyphosphorylmethyl 4-methylbenzenesulfonate SMILES: CCOP(=O)(COS(=O)(=O)C1=CC=C(C=C1)C)OCC