missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (p-Toluenesulfonyloxymethyl)phosphonate 97.0+%, TCI America™

Catalog No. D496825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D496825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D496825G Supplier TCI America Supplier No. D496825G
Only null left

Chemical Identifiers

CAS 31618-90-3
Molecular Formula C12H19O6PS
Molecular Weight (g/mol) 322.312
MDL Number MFCD03412078
InChI Key UOEFFQWLRUBDME-UHFFFAOYSA-N
Synonym (p-Toluenesulfonyloxymethyl)phosphonic Acid Diethyl Ester, (Diethoxyphosphoryl)methyl p-Toluenesulfonate, p-Toluenesulfonic Acid (Diethoxyphosphoryl)methyl Ester
PubChem CID 4193599
IUPAC Name diethoxyphosphorylmethyl 4-methylbenzenesulfonate
SMILES CCOP(=O)(COS(=O)(=O)C1=CC=C(C=C1)C)OCC

Specifications

Color Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 322.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (p-Toluenesulfonyloxymethyl)phosphonate
TSCA No
Recommended Storage Refrigerator