accessibility menu, dialog, popup

UserName

CH3HOHNNOSOOOH(E/Z)

CAS RN 3147-14-6

IUPAC Name: 4-[2-(2-hydroxy-5-methylphenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-1-sulfonic acid
Synonyms: calmagite
Molecular Weight (g/mol): 358.37
Molecular Formula: C17H14N2O5S
InChi Key: ASFVMSDYPYMUFL-UHFFFAOYSA-N
SMILES: CC1=CC(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
Grade 
  • (4)
  • (2)
  • (1)
  • (5)
  • (3)
Molecular Weight (g/mol) 
  • (10)
  • (2)
  • (2)
Color 
  • (4)
  • (4)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (5)
  • (3)
  • (10)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)