missing translation for 'onlineSavingsMsg'
Learn More

Calmagite [Metal indicator for Ca, Mg etc.], TCI America™

Catalog No. C00081G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C00081G Supplier TCI America Supplier No. C00081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3147-14-6
Molecular Formula C17H14N2O5S
Molecular Weight (g/mol) 358.368
MDL Number MFCD00011656
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
Synonym calmagite, calmagite, indicator grade, 1-naphthalenesulfonic acid, 3-hydroxy-4-2-hydroxy-5-methylphenyl azo, 3-hydroxy-4-6-hydroxy-m-tolyl azo-1-naphthalenesulfonic acid, calmagite indicator grade, 1-naphthalenesulfonic acid, 3-hydroxy-4-2-2-hydroxy-5-methylphenyl diazenyl, vbrnloqcbcpphl-uhfffaoysa-n, calmagite metalindicatorforca,mgetc., 3-hydroxy-4-2-hydroxy-5-methyl-phenylazo-1-naphthalenesulfonic acid, e-3-hydroxy-4-2-hydroxy-5-methylphenyl diazenyl naphthalene-1-sulfonic acid
PubChem CID 6364506
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O

Specifications

Melting Point 330°C
Quantity 1 g
Formula Weight 358.37
Physical Form Crystalline Powder
Chemical Name or Material Calmagite [Metal indicator for Ca, Mg etc.]