missing translation for 'onlineSavingsMsg'
Learn More

Calmagite Indicator, 0.1% (w/v), Ricca Chemical

Catalog No. 183016
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
183016 500 mL Natural Poly Bottle
18304 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 183016 Supplier Ricca Chemical Company Supplier No. 183016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3147-14-6 , 7732-18-5
Molecular Formula C17H14N2O5S
Molecular Weight (g/mol) Mixture
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
PubChem CID 6364506
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O

Specifications

CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Concentration 0.1% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Red
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.1% W/V
Grade Indicator
Chemical Name or Material Calmagite Indicator
Show More Show Less