missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calmagite, indicator grade

Catalog No. AAB2148222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2148222 100 g
AAB2148206 5 g
AAB2148214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2148222 Supplier Thermo Scientific Chemicals Supplier No. B2148222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3147-14-6 | C17H14N2O5S | 358.368 g/mol

Calmagite is widely used in analytical chemistry as an indicator in the titration of calcium or magnesium with ethylenediaminetetraacetic acid (EDTA). It is also useful in the determination of magnesium in biological materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calmagite is widely used in analytical chemistry as an indicator in the titration of calcium or magnesium with ethylenediaminetetraacetic acid (EDTA). It is also useful in the determination of magnesium in biological materials.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 3147-14-6
Molecular Formula C17H14N2O5S
Molecular Weight (g/mol) 358.368
MDL Number MFCD00011656
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
Synonym 3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)-1-naphthalenesulfonic acid
PubChem CID 6364506
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O

Specifications

Melting Point >330°C
Beilstein 708596
Merck Index 14,1718
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 358.37
Grade Indicator
Chemical Name or Material Calmagite
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-563-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only