accessibility menu, dialog, popup

UserName

CAS RN 31253-08-4

H3COOCH3I

CAS RN 31253-08-4

IUPAC Name: ethyl 2-iodopropanoate
Synonyms: ethyl 2-iodopropanoate
Molecular Weight (g/mol): 228.03
Molecular Formula: C5H9IO2
InChi Key: AVMMXNKUHBWIMU-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)I
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results