missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Iodopropionate 97.0+%, TCI America™

Catalog No. E11985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11985G 5 g
E119825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11985G Supplier TCI America Supplier No. E11985G
Only null left

Chemical Identifiers

CAS 31253-08-4
Molecular Formula C5H9IO2
Molecular Weight (g/mol) 228.029
MDL Number MFCD02258628
InChI Key AVMMXNKUHBWIMU-UHFFFAOYSA-N
Synonym 2-Iodopropionic Acid Ethyl Ester
PubChem CID 10922254
IUPAC Name ethyl 2-iodopropanoate
SMILES CCOC(=O)C(C)I

Specifications

Boiling Point 79°C
Quantity 5 g
Formula Weight 228.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Iodopropionate
TSCA No
Recommended Storage Freezer