accessibility menu, dialog, popup

UserName

CAS RN 31230-17-8

IUPAC Name: 5-methyl-1H-pyrazol-3-amine
Synonyms: 5-methyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 97.12
Molecular Formula: C4H7N3
InChi Key: FYTLHYRDGXRYEY-UHFFFAOYSA-N
SMILES: CC1=CC(N)=NN1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (5)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (4)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)