accessibility menu, dialog, popup

UserName

CAS RN 312-30-1

IUPAC Name: 1-fluoro-4-(4-fluoro-3-nitrobenzenesulfonyl)-2-nitrobenzene
Synonyms: difluorodinitrobenzene sulfone
Molecular Weight (g/mol): 344.24
Molecular Formula: C12H6F2N2O6S
InChi Key: KHAWDEWNXJIVCJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1F)S(=O)(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results