missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-fluoro-3-nitrophenyl) Sulfone 98.0+%, TCI America™

Catalog No. D053610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D053610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D053610G Supplier TCI America Supplier No. D053610G

Chemical Identifiers

CAS 312-30-1
Molecular Formula C12H6F2N2O6S
Molecular Weight (g/mol) 344.245
MDL Number MFCD00007057
InChI Key KHAWDEWNXJIVCJ-UHFFFAOYSA-N
Synonym bis 4-fluoro-3-nitrophenyl sulfone, 4-fluoro-3-nitrophenyl sulfone, difluorodinitrobenzene sulfone, sulfone, bis 4-fluoro-3-nitrophenyl, p,p'-difluoro-m,m'-dinitrodiphenyl sulfone, 3,3'-dinitro-4,4'-difluorodiphenyl sulfone, benzene, 1,1'-sulfonylbis 4-fluoro-3-nitro, bis 4-fluoro-3-nitrophenyl sulphone, nds
PubChem CID 9398
IUPAC Name 1-fluoro-4-(4-fluoro-3-nitrophenyl)sulfonyl-2-nitrobenzene
SMILES C1=CC(=C(C=C1S(=O)(=O)C2=CC(=C(C=C2)F)[N+](=O)[O-])[N+](=O)[O-])F

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 10 g
Formula Weight 344.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-fluoro-3-nitrophenyl) Sulfone
RTECSNumber WR4300000
TSCA Yes