accessibility menu, dialog, popup

UserName

CAS RN 3119-93-5

IUPAC Name: 3-ethyl-2-methyl-1,3-benzothiazol-3-ium iodide
Synonyms: 3-ethyl-2-methyl-1,3-benzothiazol-3-ium iodide
Molecular Weight (g/mol): 305.18
Molecular Formula: C10H12INS
InChi Key: HWFCSPDBFXYFKY-UHFFFAOYSA-M
SMILES: [I-].CC[N+]1=C(C)SC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)