missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-methylbenzothiazolium Iodide 98.0+%, TCI America™

Catalog No. E04765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04765G 5 g
E047625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04765G Supplier TCI America Supplier No. E04765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3119-93-5
Molecular Formula C10H12INS
Molecular Weight (g/mol) 305.177
MDL Number MFCD00044134
InChI Key HWFCSPDBFXYFKY-UHFFFAOYSA-M
PubChem CID 2729630
IUPAC Name 3-ethyl-2-methyl-1,3-benzothiazol-3-ium;iodide
SMILES CC[N+]1=C(SC2=CC=CC=C21)C.[I-]

Specifications

Melting Point 196°C
Color Green-White
Quantity 5 g
Formula Weight 305.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl-2-methylbenzothiazolium Iodide