accessibility menu, dialog, popup

UserName

CAS RN 31037-02-2

IUPAC Name: ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
Synonyms: ethyl 5-amino-1-methylpyrazole-4-carboxylate
Molecular Weight (g/mol): 169.18
Molecular Formula: C7H11N3O2
InChi Key: MEUSJJFWVKBUFP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)N(C)N=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)