missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Amino-1-methylpyrazole-4-carboxylate 98.0+%, TCI America™

Catalog No. E09865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09865G 5 g
E098625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09865G Supplier TCI America Supplier No. E09865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31037-02-2
Molecular Formula C7H11N3O2
Molecular Weight (g/mol) 169.18
MDL Number MFCD00051652
InChI Key MEUSJJFWVKBUFP-UHFFFAOYSA-N
Synonym ethyl 5-amino-1-methyl-1h-pyrazole-4-carboxylate, 5-amino-1-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl5-amino-1-methylpyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid, 5-amino-1-methyl-, ethyl ester, 5-amino-1-methylpyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-1-methyl-4-pyrazolecarboxylate, ethyl 5-amino-1-methyl-pyrazole-4-carboxylate, acmc-1agpk, maybridge1_003833, ethyl-5-amino-1-methyl-1h-pyrazol-4-carboxylat
PubChem CID 271299
IUPAC Name ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(N)N(C)N=C1

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 169.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5-Amino-1-methylpyrazole-4-carboxylate
EINECSNumber (5)-6164
TSCA No