accessibility menu, dialog, popup

UserName

FHNOHO

CAS RN 3093-97-8

IUPAC Name: 6-fluoro-1H-indole-2-carboxylic acid
Synonyms: 6-fluoro-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 179.15
Molecular Formula: C9H6FNO2
InChi Key: LRTIKMXIKAOCDM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=C(F)C=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results