accessibility menu, dialog, popup

UserName

CH3NHO

CAS RN 3085-54-9

IUPAC Name: N-(4-methylphenyl)formamide
Synonyms: P-methylformanilide
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: GRVKDWHXLFEVBP-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results