missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylformanilide 98.0+%, TCI America™

Catalog No. M17215G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17215G Supplier TCI America Supplier No. M17215G

Chemical Identifiers

CAS 3085-54-9
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00014124
InChI Key GRVKDWHXLFEVBP-UHFFFAOYSA-N
Synonym N-Formyl-p-toluidine, N-(p-Tolyl)formamide
PubChem CID 76519
IUPAC Name N-(4-methylphenyl)formamide
SMILES CC1=CC=C(NC=O)C=C1

Specifications

Melting Point 53°C
Quantity 5 g
Formula Weight 135.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Methylformanilide