accessibility menu, dialog, popup

UserName

H3COOOClClCl

CAS RN 30668-97-4

IUPAC Name: 2-ethoxyethyl 2,2,2-trichloroacetate
Synonyms: 2-ethoxyethyl 2,2,2-trichloroacetate
Molecular Weight (g/mol): 235.49
Molecular Formula: C6H9Cl3O3
InChi Key: LEGOWVFXOHMHKG-UHFFFAOYSA-N
SMILES: CCOCCOC(=O)C(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results