missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl Trichloroacetate 95.0+%, TCI America™

Catalog No. T147425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T147425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T147425G Supplier TCI America Supplier No. T147425G
Only null left

Chemical Identifiers

CAS 30668-97-4
Molecular Formula C6H9Cl3O3
Molecular Weight (g/mol) 235.49
MDL Number MFCD00191691
InChI Key LEGOWVFXOHMHKG-UHFFFAOYSA-N
Synonym Trichloroacetic Acid 2-Ethoxyethyl Ester, Ethylene Glycol Monoethyl Ether Trichloroacetate
PubChem CID 547792
IUPAC Name 2-ethoxyethyl 2,2,2-trichloroacetate
SMILES CCOCCOC(=O)C(Cl)(Cl)Cl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 235.49
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethyl Trichloroacetate