accessibility menu, dialog, popup

UserName

CAS RN 3052-61-7

H3CNH3CSS

CAS RN 3052-61-7

IUPAC Name: N,N-diethyl(benzylsulfanyl)carbothioamide
Synonyms: N,N-diethylbenzylsulfanylcarbothioamide
Molecular Weight (g/mol): 239.4
Molecular Formula: C12H17NS2
InChi Key: SNBMGLUIIGFNFE-UHFFFAOYSA-N
SMILES: CCN(CC)C(=S)SCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results