missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Diethyldithiocarbamate 98.0+%, TCI America™

Catalog No. D23135G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23135G Supplier TCI America Supplier No. D23135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3052-61-7
Molecular Formula C12H17NS2
Molecular Weight (g/mol) 239.395
MDL Number MFCD00191425
InChI Key SNBMGLUIIGFNFE-UHFFFAOYSA-N
Synonym Diethyldithiocarbamic Acid Benzyl Ester
PubChem CID 590455
IUPAC Name benzyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SCC1=CC=CC=C1

Specifications

Boiling Point 172°C
Quantity 5 g
Formula Weight 239.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Diethyldithiocarbamate