CAS RN 30483-75-1
IUPAC Name:
4-(4-bromophenyl)morpholine
Synonyms:
4-(4-bromophenyl)morpholine
Molecular Weight (g/mol):
242.12
Molecular Formula:
C10H12BrNO
InChi Key:
UJTKZWNRUPTHSB-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)N1CCOCC1
1
–
1
of
1
results
1
4-(4-Bromophenyl)morpholine 98.0+%, TCI America™
CAS: 30483-75-1 Molecular Formula: C10H12BrNO Molecular Weight (g/mol): 242.116 MDL Number: MFCD04112483 InChI Key: UJTKZWNRUPTHSB-UHFFFAOYSA-N PubChem CID: 11622851 IUPAC Name: 4-(4-bromophenyl)morpholine SMILES: C1COCCN1C2=CC=C(C=C2)Br