accessibility menu, dialog, popup

UserName

CH3SBr

CAS RN 30318-99-1

IUPAC Name: 3-bromo-4-methylthiophene
Synonyms: 3-bromo-4-methylthiophene
Molecular Weight (g/mol): 177.06
Molecular Formula: C5H5BrS
InChi Key: MBUSOPVRLCFJCS-UHFFFAOYSA-N
SMILES: CC1=CSC=C1Br
Boiling Point 
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)