missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylthiophene 93.0+%, TCI America™

Catalog No. B15415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15415G 5 g
B154125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15415G Supplier TCI America Supplier No. B15415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30318-99-1
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.059
MDL Number MFCD00130084
InChI Key MBUSOPVRLCFJCS-UHFFFAOYSA-N
Synonym 3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene
PubChem CID 2734935
IUPAC Name 3-bromo-4-methylthiophene
SMILES CC1=CSC=C1Br

Specifications

Color Yellow
Boiling Point 180°C
Quantity 5 g
Formula Weight 177.06
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-methylthiophene