accessibility menu, dialog, popup

UserName

CH3NFNH2

CAS RN 301222-66-2

IUPAC Name: 5-fluoro-4-methylpyridin-2-amine
Synonyms: 5-fluoro-4-methylpyridin-2-amine
Molecular Weight (g/mol): 126.13
Molecular Formula: C6H7FN2
InChi Key: RKMOPXJNMQWPLH-UHFFFAOYSA-N
SMILES: CC1=CC(N)=NC=C1F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)