Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-fluoro-4-methylpyridine, 95%

Catalog No. AAH6481506
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6481506 5 g
AAH6481503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH6481506 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H6481506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 301222-66-2 | C6H7FN2 | 126.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 301222-66-2
Molecular Formula C6H7FN2
Molecular Weight (g/mol) 126.134
MDL Number MFCD06659479
InChI Key RKMOPXJNMQWPLH-UHFFFAOYSA-N
Synonym 2-amino-5-fluoro-4-picoline, 2-amino-5-fluoro-4-methylpyridine, 5-fluoro-4-methyl-2-pyridinamine, 2-amino-4-methyl-5-fluoropyridine, pubchem1256, 5-fluoro-4-methyl-pyridin-2-amine, 2-pyridinamine,5-fluoro-4-methyl, 2-amino-4-methyl-5-fluoro pyridine, e 5-fluoro-4-methyl-2-pyridinamine, 2-pyridinamine, 5-fluoro-4-methyl-9ci
PubChem CID 22063670
IUPAC Name 5-fluoro-4-methylpyridin-2-amine
SMILES CC1=CC(=NC=C1F)N

Specifications

Quantity 5 g
Beilstein 9694924
Formula Weight 126.13
Percent Purity 95%
Chemical Name or Material 2-Amino-5-fluoro-4-methylpyridine
missing translation for 'healthHazard1' H302+H312+H332-H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only