accessibility menu, dialog, popup

UserName

CAS RN 3011-34-5

IUPAC Name: 4-hydroxy-3-nitrobenzaldehyde
Synonyms: 4-formyl-2-nitrophenol
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: YTHJCZRFJGXPTL-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=O)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)