missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrobenzaldehyde 97.0+%, TCI America™

Catalog No. H09245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09245G 5 g
H092425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09245G Supplier TCI America Supplier No. H09245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3011-34-5
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007117
InChI Key YTHJCZRFJGXPTL-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrobenzaldehyde, benzaldehyde, 4-hydroxy-3-nitro, 3-nitro-4-hydroxybenzaldehyde, 4-formyl-2-nitrophenol, 4-hydroxy-3-nitro-benzaldehyde, pubchem8135, acmc-1cjnu, d03vzf, 4-hydroxy-3-nitrobenzaldeyde, ksc223k5f
PubChem CID 18169
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])O

Specifications

Melting Point 143°C
Quantity 5 g
Formula Weight 167.12
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-nitrobenzaldehyde