accessibility menu, dialog, popup

UserName

CAS RN 2998-56-3

IUPAC Name: N,N-bis(2-chloroethyl)carbamoyl chloride
Synonyms: N,N-bis(2-chloroethyl)carbamoyl chloride
Molecular Weight (g/mol): 204.48
Molecular Formula: C5H8Cl3NO
InChi Key: JAHXVUPWHXMPLG-UHFFFAOYSA-N
SMILES: ClCCN(CCCl)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results