missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-chloroethyl)carbamoyl Chloride 97.0+%, TCI America™

Catalog No. B14511G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14511G Supplier TCI America Supplier No. B14511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2998-56-3
Molecular Formula C5H8Cl3NO
Molecular Weight (g/mol) 204.48
MDL Number MFCD00144974
InChI Key JAHXVUPWHXMPLG-UHFFFAOYSA-N
PubChem CID 76346
IUPAC Name N,N-bis(2-chloroethyl)carbamoyl chloride
SMILES ClCCN(CCCl)C(Cl)=O

Specifications

Color Yellow
Boiling Point 125°C
Quantity 1 g
UN Number 3265
Formula Weight 204.48
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Bis(2-chloroethyl)carbamoyl Chloride
RTECSNumber FD2200000
TSCA No