accessibility menu, dialog, popup

UserName

N

CAS RN 2981-10-4

IUPAC Name: 1-(cyclohex-1-en-1-yl)piperidine
Synonyms: 1-(cyclohex-1-en-1-yl)piperidine
Molecular Weight (g/mol): 165.28
Molecular Formula: C11H19N
InChi Key: KPVMGWQGPJULFL-UHFFFAOYSA-N
SMILES: C1CCN(CC1)C1=CCCCC1
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)