accessibility menu, dialog, popup

UserName

ClNNNN

CAS RN 298-96-4

IUPAC Name: triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: tetrazolium chloride
Molecular Weight (g/mol): 334.81
Molecular Formula: C19H15ClN4
InChi Key: PKDBCJSWQUOKDO-UHFFFAOYSA-M
SMILES: [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (14)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (4)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (4)
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (4)
  • (2)
  • (2)
  • (5)
  • (4)
  • (2)
  • (3)
  • (4)
  • (14)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (6)