missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triphenyltetrazolium Chloride, Reagents

Catalog No. 01336904
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336904 100 g
01336903 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336904 Supplier Labchem Inc Supplier No. C2342700100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2,3,5-Triphenyltetrazolium Chloride, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 298-96-4
Molecular Formula C19H15ClN4
Molecular Weight (g/mol) 334.81
InChI Key PKDBCJSWQUOKDO-UHFFFAOYSA-M
IUPAC Name triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White to Yellow
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 1
Grade Reagent
Physical Form Solid
Chemical Name or Material 2,3,5-Triphenyltetrazolium Chloride