accessibility menu, dialog, popup

UserName

CAS RN 29799-07-3

IUPAC Name: 4-(adamantan-1-yl)phenol
Synonyms: 4-(adamantan-1-yl)phenol
Molecular Weight (g/mol): 228.34
Molecular Formula: C16H20O
InChi Key: KZMYFIUFUAOZHP-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results