missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Adamantyl)phenol 99.0+%, TCI America™

Catalog No. A13925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13925G 5 g
A13921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13925G Supplier TCI America Supplier No. A13925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29799-07-3
Molecular Formula C16H20O
Molecular Weight (g/mol) 228.335
MDL Number MFCD00168143
InChI Key KZMYFIUFUAOZHP-UHFFFAOYSA-N
Synonym 1-(4-Hydroxyphenyl)adamantane
PubChem CID 269933
IUPAC Name 4-(1-adamantyl)phenol
SMILES C1C2CC3CC1CC(C2)(C3)C4=CC=C(C=C4)O

Specifications

Melting Point 179°C
Color White
Quantity 5 g
Formula Weight 228.34
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Adamantyl)phenol