accessibility menu, dialog, popup

UserName

NH2SHOON

CAS RN 29676-71-9

IUPAC Name: 2-(2-amino-1,3-thiazol-4-yl)acetic acid
Synonyms: (2-amino-1,3-thiazol-4-yl)acetic acid
Molecular Weight (g/mol): 158.18
Molecular Formula: C5H6N2O2S
InChi Key: DYCLHZPOADTVKK-UHFFFAOYSA-N
SMILES: NC1=NC(CC(O)=O)=CS1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)