missing translation for 'onlineSavingsMsg'
Learn More

(2-Amino-4-thiazolyl)acetic Acid 98.0+%, TCI America™

Catalog No. A095425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A095425G 25 g
A0954250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A095425G Supplier TCI America Supplier No. A095425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29676-71-9
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.175
MDL Number MFCD00010068
InChI Key DYCLHZPOADTVKK-UHFFFAOYSA-N
Synonym 2-amino-4-thiazoleacetic acid, 2-2-aminothiazol-4-yl acetic acid, 2-aminothiazol-4-acetic acid, 2-aminothiazole-4-yl acetic acid, 4-thiazoleacetic acid, 2-amino, 2-amino-1,3-thiazol-4-yl acetic acid, 2-aminothiazole-4-yl-acetic acid, 2-aminothiazole-4-acetic acid, unii-qsh3o4tnjt, 2-2-amino-1,3-thiazol-4-yl acetic acid
PubChem CID 34665
IUPAC Name 2-(2-amino-1,3-thiazol-4-yl)acetic acid
SMILES C1=C(N=C(S1)N)CC(=O)O

Specifications

Melting Point 123°C
Quantity 25 g
Formula Weight 158.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Amino-4-thiazolyl)acetic Acid
TSCA No
Recommended Storage Refrigerator