accessibility menu, dialog, popup

UserName

CAS RN 2942-58-7

CH3OPOOCH3N

CAS RN 2942-58-7

IUPAC Name: diethyl cyanophosphonate
Synonyms: diethylphosphorocyanidate
Molecular Weight (g/mol): 163.11
Molecular Formula: C5H10NO3P
InChi Key: ZWWWLCMDTZFSOO-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results