CAS RN 29335-36-2
IUPAC Name:
N'-hydroxypropanimidamide
Synonyms:
N'-hydroxypropanimidamide
Molecular Weight (g/mol):
88.11
Molecular Formula:
C3H8N2O
InChi Key:
RLZPCFQNZGINRP-UHFFFAOYSA-N
SMILES:
CCC(N)=NO
1
–
1
of
1
results
1
Promotions Available
Propionamidoxime, 97%
Propionamidoxime, 97%
CAS: 29335-36-2 Molecular Formula: C3H8N2O Molecular Weight (g/mol): 88.11 MDL Number: MFCD03426272 InChI Key: RLZPCFQNZGINRP-UHFFFAOYSA-N Synonym: n-hydroxypropionamidine,n-hydroxypropionimidamide,1z-n'-hydroxypropanimidamide,1-hydroxyimino propylamine,n-hydroxy-propionamidine,n-hydroxyproponmdamde,n-hydroxypropanamidine,n'-hydroxypropimidamide,z-n'-hydroxypropimidamide,e-n'-hydroxypropanimidamide PubChem CID: 6521662 IUPAC Name: N'-hydroxypropanimidamide SMILES: CCC(=NO)N