missing translation for 'onlineSavingsMsg'
Learn More

Propionamidoxime, 97%

Catalog No. AAH50889MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50889MD 250 mg
AAH5088903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50889MD Supplier Thermo Scientific Chemicals Supplier No. H50889MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29335-36-2 | C3H8N2O | 88.11 g/mol

Propionamidoxime is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propionamidoxime is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 29335-36-2
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD03426272
InChI Key RLZPCFQNZGINRP-UHFFFAOYSA-N
Synonym n-hydroxypropionamidine, n-hydroxypropionimidamide, 1z-n'-hydroxypropanimidamide, 1-hydroxyimino propylamine, n-hydroxy-propionamidine, n-hydroxyproponmdamde, n-hydroxypropanamidine, n'-hydroxypropimidamide, z-n'-hydroxypropimidamide, e-n'-hydroxypropanimidamide
PubChem CID 6521662
IUPAC Name N'-hydroxypropanimidamide
SMILES CCC(=NO)N

Specifications

Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 88.11
Percent Purity 97%
Chemical Name or Material Propionamidoxime
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only