accessibility menu, dialog, popup

UserName

OONClBr

CAS RN 29241-62-1

IUPAC Name: 5-bromo-6-chloropyridine-3-carboxylate
Synonyms: 5-bromo-6-chloropyridine-3-carboxylate
Molecular Weight (g/mol): 235.44
Molecular Formula: C6H2BrClNO2
InChi Key: DXEUARPQHJXMII-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CN=C(Cl)C(Br)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)