missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-6-chloronicotinic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH3238406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3238406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3238406 Supplier Thermo Scientific Chemicals Supplier No. H3238406

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29241-62-1
Molecular Formula C6H2BrClNO2
Molecular Weight (g/mol) 235.44
MDL Number MFCD01927098
InChI Key DXEUARPQHJXMII-UHFFFAOYSA-M
Synonym 5-bromo-6-chloronicotinic acid, 5-bromo-6-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 5-bromo-6-chloro, 3-bromo-2-chloropyridine-5-carboxylic acid, 3-bromo-2-chloro-5-pyridinecarboxylic acid, zlchem 1120, pubchem12922, acmc-1ckhn, ksc203c5j, abbypharma ap-12-5123
PubChem CID 285433
SMILES [O-]C(=O)C1=CN=C(Cl)C(Br)=C1

Specifications

Melting Point 170°C to 173°C
Quantity 5 g
IUPAC Name 5-bromo-6-chloropyridine-3-carboxylic acid
Formula Weight 236.45
Percent Purity 97%
Chemical Name or Material 5-Bromo-6-chloronicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only